Learn More
Cayman Chemical 6-MethylmercaptopurIn-d3 10mg
SDP
Catalog No. 502912602
Encompass_Preferred
An internal standard for the quantification of 6-MMP by GC- or LC-MS
Catalog No. 50-291-2602 Supplier Cayman Chemical Supplier No. 3632110 MG
An internal standard for the quantification of 6-MMP by GC- or LC-MS