Learn More
Cayman Chemical 6-Methylmercaptopurine-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 33312-93-5, 25 mg
SDP
Catalog No. 502912603
Encompass_Preferred
An internal standard for the quantification of 6-MMP by GC- or LC-MS
Catalog No. 50-291-2603 Supplier Cayman Chemical Supplier No. 3632125 MG
An internal standard for the quantification of 6-MMP by GC- or LC-MS