Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-Methylpicolinic acid | 934-60-1 | 137.14 | 1 ML
SDP

Catalog No. 5000447158
Encompass_Preferred

6-Methylpicolinic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only and not for patient use.

  • Solid appearance: off-white to light brown
  • Solubility in DMSO: 100 mg/mL (729.18 mM), requiring ultrasonic agitation
  • Solubility in vivo: forms clear solutions at ≥ 2.5 mg/mL (18.23 mM) in various protocols (e.g., 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline)
  • Storage conditions for powder: -20°C for 3 years; 4°C for 2 years
  • Storage conditions for in solvent/stock solution: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-47158 Supplier Medchemexpress LLC Supplier No. HYW00213910MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-Methylpicolinic acid is a biochemical reagent used as a biological material or organic compound for life science related research. It is intended for research use only and not for patient use.

  • Solid appearance: off-white to light brown
  • Solubility in DMSO: 100 mg/mL (729.18 mM), requiring ultrasonic agitation
  • Solubility in vivo: forms clear solutions at ≥ 2.5 mg/mL (18.23 mM) in various protocols (e.g., 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline)
  • Storage conditions for powder: -20°C for 3 years; 4°C for 2 years
  • Storage conditions for in solvent/stock solution: -80°C for 6 months; -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.