Learn More
Medchemexpress LLC 6-Methylpicolinic acid | 934-60-1 | 137.14 | 25 G
SDP
Catalog No. 5000448108
Encompass_Preferred
6-Methylpicolinic acid (6-Methylpyridine-2-carboxylic acid) is a biochemical reagent for life science research. It is an organic compound and biological material, intended for research use only. The product is an off-white to light brown solid.
  • Biochemical reagent for life science research.
  • Functions as an organic compound and biological material.
  • High purity of 99.82% and 99.85%.
  • Off-white to light brown solid appearance.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Solution storage: -80°C for 6 months; -20°C for 1 month.
  • Specific protocols for in vivo dissolution available.
  • Freshly prepared working solutions recommended for in vivo experiments.
  • Supporting documentation (Data Sheet, COA, SDS, HNMR, RP-HPLC, Handling Instructions) available.
Catalog No. 50-004-48108 Supplier Medchemexpress LLC Supplier No. 25GHYW002139
6-Methylpicolinic acid (6-Methylpyridine-2-carboxylic acid) is a biochemical reagent for life science research. It is an organic compound and biological material, intended for research use only. The product is an off-white to light brown solid.
  • Biochemical reagent for life science research.
  • Functions as an organic compound and biological material.
  • High purity of 99.82% and 99.85%.
  • Off-white to light brown solid appearance.
  • Soluble in DMSO at 100 mg/mL.
  • Powder storage: -20°C for 3 years; 4°C for 2 years.
  • Solution storage: -80°C for 6 months; -20°C for 1 month.
  • Specific protocols for in vivo dissolution available.
  • Freshly prepared working solutions recommended for in vivo experiments.
  • Supporting documentation (Data Sheet, COA, SDS, HNMR, RP-HPLC, Handling Instructions) available.