Learn More
Cayman Chemical 6-MethylpterIn 100mg
SDP
Catalog No. 502913390
Encompass_Preferred
A derivative of folic acid; generates singlet oxygen and hydrogen peroxide when exposed to photoirradiation in cell-free assays; has been used as an internal standard for the detection of pterins in urine
Catalog No. 50-291-3390 Supplier Cayman Chemical Supplier No. 35380100 MG
A derivative of folic acid; generates singlet oxygen and hydrogen peroxide when exposed to photoirradiation in cell-free assays; has been used as an internal standard for the detection of pterins in urine