Learn More
Cayman Chemical 6-Methylpterin, Purity: ≥98%, CAS: 708-75-8, 50 mg
SDP
Catalog No. 502913640
Encompass_Preferred
A derivative of folic acid; generates singlet oxygen and hydrogen peroxide when exposed to photoirradiation in cell-free assays; has been used as an internal standard for the detection of pterins in urine
Catalog No. 50-291-3640 Supplier Cayman Chemical Supplier No. 3538050 MG
A derivative of folic acid; generates singlet oxygen and hydrogen peroxide when exposed to photoirradiation in cell-free assays; has been used as an internal standard for the detection of pterins in urine