Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylpurine, 98%, Thermo Scientific™

Catalog No. AC226572500
Change view
Click to view available options
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC226572500 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC226572500 Supplier Thermo Scientific Chemicals Supplier No. 226572500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2004-03-7
CAS Min % 97.5
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C6H6N4
Quantity 250mg
MDL Number MFCD00083663
Beilstein 26, 358
Synonym 6-methylpurine, 6-methyl-7h-purine, 6-methyl-9h-purine, 1h-purine, 6-methyl, 6-methyl-1h-purine, purine, 6-methyl, 6-methyl purine, 1h-purine, 6-methyl-9ci, symhuefssmbhja-uhfffaoysa-n, 1h-purine,6-methyl
InChI Key SYMHUEFSSMBHJA-UHFFFAOYSA-N
Molecular Weight (g/mol) 134.14
PubChem CID 5287547
ChEBI CHEBI:40274
Formula Weight 134.14
Percent Purity ≥97.5%
Chemical Name or Material 6-Methylpurine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Toxic if inhaled.
May cause an allergic skin reaction.
Toxic if swallowed.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality
EINECSNumber 217-903-2
RTECSNumber UO8700000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.