Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Methylquinoline

Catalog No. 10892825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-892-825G 25 g
10-892-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-892-825G Supplier Sigma Aldrich Supplier No. 10892825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 6-methylquinoline can be used as primary carbon source in culture of pseudomonas putida qp1
  • 6-methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.6-methylquinoline undergoes biodegradation by quinoline-degrading culture of pseudomonas putida

Specifications

CAS 91-62-3
Density 1.067 g/mL (at 20°C (literature))
Boiling Point 256°C to 260°C (lit.)
Molecular Formula C10H9N
Refractive Index n20/D 1.614 (literature)
Linear Formula C10H9N
MDL Number MFCD00006804
Quantity 25 g
Synonym p-Toluquinoline; NSC 4152
Molecular Weight (g/mol) 143.19
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.