Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylquinoline, 98%, Thermo Scientific Chemicals

Catalog No. AC295141000
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC295141000 100 g
AC295140100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC295141000 Supplier Thermo Scientific Chemicals Supplier No. 295141000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 91-62-3 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-62-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006804
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
PubChem CID 7059
IUPAC Name 6-methylquinoline
SMILES CC1=CC=C2N=CC=CC2=C1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Density 1.0600g/mL
Boiling Point 256.0°C to 260.0°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.6125 to 1.6145
Quantity 100 g
Specific Gravity 1.06
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
Formula Weight 143.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 6-Methylquinoline, 95%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before re
EINECSNumber 202-084-6
RTECSNumber VC0550000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.