Learn More
Abcam 6-(Methylsulfinyl)he x yl isothiocyanate (6-MITC), tumor cell proliferation inhibitor, 25MG
Catalog No. 502706153
Encompass_Preferred
MW 205.3 Da, Purity >98%. Potent tumor cell proliferation inhibitor in vitro (IC₅₀ = 8 μM). Platelet aggregation inhibitor. Anti-inflammatory and anticancer agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6153 Supplier Abcam Supplier No. AB14289825MG
MW 205.3 Da, Purity >98%. Potent tumor cell proliferation inhibitor in vitro (IC₅₀ = 8 μM). Platelet aggregation inhibitor. Anti-inflammatory and anticancer agent. Active in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement