Learn More
Medchemexpress LLC 6-Methyluridine | 16710-13-7 | C10H14N2O6 | 100 MG
SDP
Catalog No. 5000373094
Encompass_Preferred
6-Methyluridine is a uridine analogue used for research purposes. This compound can be utilized to study anticonvulsant and anxiolytic activities, and in the development of new antihypertensive agents. It is strictly for research use.
  • Uridine analogue
  • Potential antiepileptic effects
  • Used to study anticonvulsant and anxiolytic activities
  • Aids in developing new antihypertensive agents
  • For research use only
Catalog No. 50-003-73094 Supplier Medchemexpress LLC Supplier No. HY152732100MG
6-Methyluridine is a uridine analogue used for research purposes. This compound can be utilized to study anticonvulsant and anxiolytic activities, and in the development of new antihypertensive agents. It is strictly for research use.
  • Uridine analogue
  • Potential antiepileptic effects
  • Used to study anticonvulsant and anxiolytic activities
  • Aids in developing new antihypertensive agents
  • For research use only