Learn More
Accela Chembio Inc 6-metyl-5-nitrouracil | 5g | 16632-21-6 | MFCD00047362 | 98% | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50250022
Encompass_Preferred
6-metyl-5-nitrouracil | 5g | 16632-21-6 | MFCD00047362 | 98% | Shelf Life: 1260 Days | Light Sensitive
Catalog No. 50-250-022 Supplier Accela Chembio Inc Supplier No. SY0030795G
6-metyl-5-nitrouracil | 5g | 16632-21-6 | MFCD00047362 | 98% | Shelf Life: 1260 Days | Light Sensitive