Learn More
Cole-Parmer Nylon Micro Brush, 3.5" Handle, 2.5"L x 0.25" Dia Brush; 12/Pk
Catalog No. NC0098540 Shop All Cole-Parmer Products
Encompass
Nylon Micro Brush, 3.5" Handle, 2.5"L x 0.25" Dia Brush; 12/PkBrush design is ideal for cleaning 2- to 100-ml centrifuge tubes, micro tubes, and a variety of other containers.
Catalog No. NC0098540 Supplier Cole-Parmer Supplier No. 8455194
Nylon Micro Brush, 3.5" Handle, 2.5"L x 0.25" Dia Brush; 12/PkBrush design is ideal for cleaning 2- to 100-ml centrifuge tubes, micro tubes, and a variety of other containers.