Learn More
Cellvis 6 MICRO-WELL GLASS BOTTOM PLAT
Catalog No. NC0454735
Encompass
6 micro-well glass bottom plate with #1.5 cover glass(0.16-0.19). Micro-well size 20mm, with lid, Individually packed. 20 per case.
Catalog No. NC0454735 Supplier Cellvis Supplier No. P06201.5N
6 micro-well glass bottom plate with #1.5 cover glass(0.16-0.19). Micro-well size 20mm, with lid, Individually packed. 20 per case.