Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Morpholino-3-pyridinyl isothiocyanate, 97%, Thermo Scientific™

Catalog No. CC17507CB
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC17507CB 250 mg
CC17507DA 1 g
CC17507DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC17507CB Supplier Thermo Scientific Chemicals Supplier No. CC17507CB
Add to Cart

Chemical Identifiers

CAS 52024-29-0
Molecular Formula C10H11N3OS
Molecular Weight (g/mol) 221.278
MDL Number MFCD03086114
InChI Key VLZWMPRZGPULIU-UHFFFAOYSA-N
Synonym 4-5-isothiocyanatopyridin-2-yl morpholine, 6-morpholino-3-pyridinyl isothiocyanate, 6-morpholino-3-pyridinylisothiocyanate, 4-5-isothiocyanato-2-pyridyl morpholine, 4-5-isothiocyatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-3-isothiocyanate, 4-5-isothiocyanato-2-pyridinyl morpholine, 5-isothiocyanato-2-morpholin-4-yl pyridine, morpholine,4-5-isothiocyanato-2-pyridinyl, 6-morpholin-4-yl pyridin-3-yl isothiocyanate
PubChem CID 2776464
IUPAC Name 4-(5-isothiocyanatopyridin-2-yl)morpholine
SMILES C1COCCN1C2=NC=C(C=C2)N=C=S

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 221.28
Percent Purity 97%
Chemical Name or Material 6-Morpholino-3-pyridinyl isothiocyanate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.