Learn More
MSC Mitutoyo 500-196-30 Absolute Digimatic Caliper, 0 to 6"/0 to 150mm Measuring Range Advanced Onsite Sensor (AOS), 0.0005"/0.01mm Resolution, LCD Stainless Steel with 40mm Stainless Steel Jaws, 0.0300mm Accuracy
Catalog No. 501113268
Encompass
Mitutoyo 500-196-30 Absolute Digimatic Caliper, 0 to 6"/0 to 150mm Measuring Range Advanced Onsite Sensor (AOS), 0.0005"/0.01mm Resolution, LCD Stainless Steel with 40mm Stainless Steel Jaws, 0.0300mm Accuracy
Catalog No. 50-111-3268 Supplier MSC Supplier No. 62529482
Mitutoyo 500-196-30 Absolute Digimatic Caliper, 0 to 6"/0 to 150mm Measuring Range Advanced Onsite Sensor (AOS), 0.0005"/0.01mm Resolution, LCD Stainless Steel with 40mm Stainless Steel Jaws, 0.0300mm Accuracy