Learn More
Medchemexpress LLC 6-(N-Boc)caproic acid N-hydroxysuccinimide ester | 51513-80-5 | MFCD00237379 | ≥98.0% | 328.36 g·mol-1 | C15H24N2O6 | 500 MG
SDP
Catalog No. 5000301973
Encompass_Preferred
6-(N-Boc)caproic acid NHS is the N-hydroxysuccinimide (NHS) ester of N-Boc-aminocaproic acid used as an amine-reactive coupling reagent and protein cross-linker for research applications. The NHS activation enables efficient conjugation to primary amines under mild conditions, while the N-Boc group provides an amine-protected spacer for selective chemistry. For research use only.
  • Activates carboxyl groups for rapid coupling to primary amines.
  • Provides a protected aminohexanoic spacer to reduce steric effects.
  • Suitable for protein, peptide, and bioconjugation workflows.
  • Available in laboratory pack sizes for flexible experimental use.
  • Supplied as a solid for convenient storage and handling.
Catalog No. 50-003-01973 Supplier Medchemexpress LLC Supplier No. HYW040113500MG
6-(N-Boc)caproic acid NHS is the N-hydroxysuccinimide (NHS) ester of N-Boc-aminocaproic acid used as an amine-reactive coupling reagent and protein cross-linker for research applications. The NHS activation enables efficient conjugation to primary amines under mild conditions, while the N-Boc group provides an amine-protected spacer for selective chemistry. For research use only.
  • Activates carboxyl groups for rapid coupling to primary amines.
  • Provides a protected aminohexanoic spacer to reduce steric effects.
  • Suitable for protein, peptide, and bioconjugation workflows.
  • Available in laboratory pack sizes for flexible experimental use.
  • Supplied as a solid for convenient storage and handling.