Learn More
Medchemexpress LLC 6-(N-Boc)caproic acid N-hydroxysuccinimide ester | 51513-80-5 | MFCD00237379 | ≥98.0% | 328.36 g·mol⁻¹ | C15H24N2O6 | 25 G
SDP
Catalog No. 5000304027
Encompass_Preferred
6-(N-Boc)caproic acid NHS is the N-hydroxysuccinimide ester of N-Boc-protected caproic acid used as an amine-reactive cross-linker for bioconjugation and peptide synthesis. Supplied as a white to off-white solid for research use, it activates carboxyl groups to enable efficient coupling to primary amines.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • N-Boc protection stabilizes the amino functionality during handling and reactions.
  • Suitable for protein cross-linking and peptide synthesis applications.
  • Solid form with recommended frozen storage for extended stability.
  • Available in multiple package sizes to support small-scale and larger preparations.
  • For research use only; not for human or clinical use.
Catalog No. 50-003-04027 Supplier Medchemexpress LLC Supplier No. 25GHYW040113
6-(N-Boc)caproic acid NHS is the N-hydroxysuccinimide ester of N-Boc-protected caproic acid used as an amine-reactive cross-linker for bioconjugation and peptide synthesis. Supplied as a white to off-white solid for research use, it activates carboxyl groups to enable efficient coupling to primary amines.
  • Amine-reactive NHS ester for efficient coupling to primary amines.
  • N-Boc protection stabilizes the amino functionality during handling and reactions.
  • Suitable for protein cross-linking and peptide synthesis applications.
  • Solid form with recommended frozen storage for extended stability.
  • Available in multiple package sizes to support small-scale and larger preparations.
  • For research use only; not for human or clinical use.