Learn More
Medchemexpress LLC 6-(N-Boc)caproic acid NHS | 51513-80-5 | MFCD00237379 | >=98.0% | 328.36 g/mol | C15H24N2O6 | 1 G
SDP
Catalog No. 5000299499
Encompass_Preferred
6-(N-Boc)caproic acid N-hydroxysuccinimide (NHS) ester is an activated carboxylic acid reagent used to introduce a Boc-protected aminohexanoyl spacer onto primary amines of peptides and proteins, facilitating amide bond formation for conjugation and cross-linking applications.
  • Activated NHS ester for rapid coupling to primary amines.
  • Provides Boc-protected aminohexanoyl spacer for controlled linkage.
  • Solid, white to off-white form for easy handling and storage.
  • Suitable for protein cross-linking and peptide conjugation workflows.
  • Available in multiple pack sizes to support different experimental scales.
Catalog No. 50-002-99499 Supplier Medchemexpress LLC Supplier No. HYW0401131G
6-(N-Boc)caproic acid N-hydroxysuccinimide (NHS) ester is an activated carboxylic acid reagent used to introduce a Boc-protected aminohexanoyl spacer onto primary amines of peptides and proteins, facilitating amide bond formation for conjugation and cross-linking applications.
  • Activated NHS ester for rapid coupling to primary amines.
  • Provides Boc-protected aminohexanoyl spacer for controlled linkage.
  • Solid, white to off-white form for easy handling and storage.
  • Suitable for protein cross-linking and peptide conjugation workflows.
  • Available in multiple pack sizes to support different experimental scales.