Learn More
Medchemexpress LLC 6-(N-Boc)caproic acid NHS | 51513-80-5 | MFCD00237379 | >=98.0% | 328.36 g·mol⁻¹ | C15H24N2O6 | 10 G
SDP
Catalog No. 5000299786
Encompass_Preferred
6-(N-Boc)caproic acid N-hydroxysuccinimide (NHS) ester is a Boc-protected caproic acid derivative bearing an activated NHS ester for efficient amide bond formation. It is supplied as a reactive cross-linker for coupling carboxyl groups to primary amines in protein conjugation and peptide synthesis, with the Boc group protecting amine functionality during multi-step syntheses.
  • Activated NHS ester for efficient amide bond formation with primary amines.
  • Boc protection preserves amine functionality during synthetic steps.
  • Suitable for protein cross-linking and bioconjugation workflows.
  • Available in multiple laboratory pack sizes for research use.
  • Includes downloadable datasheet and safety data sheet for handling and characterization.
Catalog No. 50-002-99786 Supplier Medchemexpress LLC Supplier No. HYW04011310G
6-(N-Boc)caproic acid N-hydroxysuccinimide (NHS) ester is a Boc-protected caproic acid derivative bearing an activated NHS ester for efficient amide bond formation. It is supplied as a reactive cross-linker for coupling carboxyl groups to primary amines in protein conjugation and peptide synthesis, with the Boc group protecting amine functionality during multi-step syntheses.
  • Activated NHS ester for efficient amide bond formation with primary amines.
  • Boc protection preserves amine functionality during synthetic steps.
  • Suitable for protein cross-linking and bioconjugation workflows.
  • Available in multiple laboratory pack sizes for research use.
  • Includes downloadable datasheet and safety data sheet for handling and characterization.