Learn More
Medchemexpress LLC 6-(N-Boc)caproic acid NHS | 51513-80-5 | MFCD00237379 | >=98.0% | 328.36 g/mol | C15H24N2O6 | 5 G
SDP
Catalog No. 5000301972
Encompass_Preferred
6-(N-Boc)caproic acid NHS is an N-hydroxysuccinimide (NHS) ester of Boc-protected caproic acid used as an amine-reactive linker and protein cross-linking reagent in bioconjugation and biochemical research. It provides an activated carboxylate for rapid coupling to primary amines and a tert-butyloxycarbonyl (Boc) protected spacer for selective deprotection after conjugation.
  • NHS-activated ester for efficient coupling to primary amines.
  • Boc-protected amino spacer enables controlled deprotection.
  • Designed for protein cross-linking and bioconjugation workflows.
  • Solid, white to off-white form for convenient handling and storage.
  • Molecular weight 328.36 g/mol; formula C15H24N2O6.
Catalog No. 50-003-01972 Supplier Medchemexpress LLC Supplier No. HYW0401135G
6-(N-Boc)caproic acid NHS is an N-hydroxysuccinimide (NHS) ester of Boc-protected caproic acid used as an amine-reactive linker and protein cross-linking reagent in bioconjugation and biochemical research. It provides an activated carboxylate for rapid coupling to primary amines and a tert-butyloxycarbonyl (Boc) protected spacer for selective deprotection after conjugation.
  • NHS-activated ester for efficient coupling to primary amines.
  • Boc-protected amino spacer enables controlled deprotection.
  • Designed for protein cross-linking and bioconjugation workflows.
  • Solid, white to off-white form for convenient handling and storage.
  • Molecular weight 328.36 g/mol; formula C15H24N2O6.