Learn More
Medchemexpress LLC 6β-naltrexol-d3 (6β-hydroxynaltrexone-d3) | 1435727-11-9 | 346.44 | C20H22D3NO4 | 1 MG
SDP
Catalog No. 5000242992
Encompass_Preferred
6β-Naltrexol-d3 (6β-hydroxynaltrexone-d3) is a deuterium-labeled stable isotope of 6β-naltrexol used as an internal standard and tracer in analytical and pharmacokinetic studies. It is the deuterated form of the primary metabolite of naltrexone and is supplied as a small-mass solid for research use (chemical formula C20H22D3NO4; MW 346.44; CAS 1435727-11-9).
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • Suitable as a tracer in pharmacokinetic and metabolic studies.
  • Represents a peripherally selective opioid antagonist metabolite.
  • Provided in small solid quantities suitable for analytical assays (e.g., 1 mg).
  • Chemical formula C20H22D3NO4 and molecular weight 346.44.
Catalog No. 50-002-42992 Supplier Medchemexpress LLC Supplier No. HY108229S1MG
6β-Naltrexol-d3 (6β-hydroxynaltrexone-d3) is a deuterium-labeled stable isotope of 6β-naltrexol used as an internal standard and tracer in analytical and pharmacokinetic studies. It is the deuterated form of the primary metabolite of naltrexone and is supplied as a small-mass solid for research use (chemical formula C20H22D3NO4; MW 346.44; CAS 1435727-11-9).
  • Deuterium-labeled internal standard for LC-MS and GC-MS.
  • Suitable as a tracer in pharmacokinetic and metabolic studies.
  • Represents a peripherally selective opioid antagonist metabolite.
  • Provided in small solid quantities suitable for analytical assays (e.g., 1 mg).
  • Chemical formula C20H22D3NO4 and molecular weight 346.44.