Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitropiperonal 97.0+%, TCI America™
SDP

Catalog No. N053725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N053725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N053725G Supplier TCI America Supplier No. N053725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 712-97-0
Molecular Formula C8H5NO5
Molecular Weight (g/mol) 195.13
MDL Number MFCD00005829
InChI Key NRZWECORTTWSEF-UHFFFAOYSA-N
Synonym 6-nitropiperonal, 6-nitrobenzo d 1,3 dioxole-5-carbaldehyde, piperonal, 6-nitro, 1,3-benzodioxole, 5-formyl-6-nitro, 2-nitro-4,5-methylenedioxybenzaldehyde, 4,5-methylenedioxy-2-nitrobenzaldehyde, 1,3-benzodioxole-5-carboxaldehyde, 6-nitro, 3,4-methylenedioxy-6-nitrobenzaldehyde, 6-nitro-1,3-benzodioxole-5-carboxaldehyde
PubChem CID 12840
IUPAC Name 6-nitro-1,3-benzodioxole-5-carbaldehyde
SMILES C1OC2=C(O1)C=C(C(=C2)C=O)[N+](=O)[O-]

Specifications

Melting Point 97°C
Color Yellow
Quantity 25 g
Formula Weight 195.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitropiperonal
RTECSNumber DF4912750
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.