Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 6-Nitroquinoline

Catalog No. N2400525G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N2400525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N2400525G Supplier Sigma Aldrich Supplier No. N2400525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Methyl 10-chloro-10-oxodecanoate (methyl sebacoyl chloride, 10-oxo-10-chlorodecanoic acid methyl ester) is a monomethyl ester of sebacinic acid.methyl 10-chloro-10-oxodecanoate may be used in the preparation of the following:
  • Methyl 10-[2-methoxy-4-(3-trifluoromethyl-3h-diazirin-3-yl)phenyl]-10-oxodecanoate
  • Fluorescein-derivatives of necrostatin-3
  • Methyl 10-oxo-10-(pyren-3-yl)decanoate

Specifications

CAS 613-50-3
Melting Point 151°C to 153°C (lit.)
Molecular Formula C9H6N2O2
Linear Formula C9H6N2O2
MDL Number MFCD00006799
Quantity 5 g
Molecular Weight (g/mol) 174.16
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.