Learn More
Medchemexpress LLC HY-N4072 1mg Medchemexpress, 6''-O-Acetylglycitin CAS:134859-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40721MG
Encompass_Preferred
Medchemexpress, HY-N4072 1mg 6''-O-Acetylglycitin CAS:134859-96-4 6''-O-Acetylglycitin, a acetyl glucoside, is one of the isoflavone isomer in soybeans, shows various extents of content reduction dependent on storage temperature, packaging condition, and its isoflavone isomer kind. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1531
 Restricted Item