Learn More
Chemglass Life Sciences Agitator, PTFE, Four Blade, 6in Diameter, Fits 25.4mm Shaft Size, 60° Impeller Angle
SDP
Catalog No. CG209517
Encompass_Preferred
Agitator, PTFE, Four Blade, 6in Diameter, Fits 25.4mm Shaft Size, 60¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 25.4mm OD stirrer shaft CG-2097 and have 60¿ impeller angles. The 60¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. PLEASE NOTE: ANGLED IMPELLARS DESIGNED TO STIR UPWARDS IN A CLOCKWISE DIRECTION ARE AVAILABLE. PLEASE CALL FOR PRICING.
Catalog No. CG209517 Supplier Chemglass Life Sciences Supplier No. CG209517
Agitator, PTFE, Four Blade, 6in Diameter, Fits 25.4mm Shaft Size, 60¿ Impeller Angle. Agitators are made entirely of PTFE and glass filled PTFE and are for use with 25.4mm OD stirrer shaft CG-2097 and have 60¿ impeller angles. The 60¿ impellers are set up to stir downwards in a clockwise direction and upwards when rotated counter-clockwise. Agitator hub is open so that it can be positioned anywhere along the shaft. Locks onto shaft with a collet hub and nut and has a glass filled PTFE safety screw. PLEASE NOTE: ANGLED IMPELLARS DESIGNED TO STIR UPWARDS IN A CLOCKWISE DIRECTION ARE AVAILABLE. PLEASE CALL FOR PRICING.