Learn More
Medchemexpress LLC 6''-O-Malonylgenistin | 51011-05-3 | 518.42 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYN09171MG
Encompass_Preferred
6''-O-Malonylgenistin, also known as Malonylgenistin, is a glycosylated isoflavone and a metabolite of Genistin. It is known for its ability to activate hTAS2R39, making it a valuable tool for research into bitter taste receptors.
  • Glycosylated isoflavone
  • Metabolite of genistin
  • Activates hTAS2R39
  • Useful for research related to bitter taste
  • Intended for scientific investigation
Catalog No. 50-004-16915