Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-methyl guanosine | 7803-88-5 | MFCD01723912 | 99.7% | 297.27 g·mol⁻1 | C11H15N5O5 | 5 MG
SDP

Catalog No. 5000263290
Encompass_Preferred

6-O-methyl guanosine is a modified nucleoside for biochemical research, reported to inhibit colony-forming ability in certain xeroderma pigmentosum cell lines. It is supplied as a solid reagent intended for in vitro studies and analytical applications.

  • High purity (99.7%) suitable for analytical and biological experiments.
  • Molecular weight 297.27 g·mol⁻1; formula C11H15N5O5.
  • Soluble in DMSO (130 mg/mL); sparingly soluble in water (2 mg/mL); ultrasonic or warming recommended to aid dissolution.
  • Stable as a powder at -20°C for extended storage.
  • Available in small research quantities (for example, 5 mg) for laboratory use.

Catalog No. 50-002-63290 Supplier Medchemexpress LLC Supplier No. HY1116485MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-methyl guanosine is a modified nucleoside for biochemical research, reported to inhibit colony-forming ability in certain xeroderma pigmentosum cell lines. It is supplied as a solid reagent intended for in vitro studies and analytical applications.

  • High purity (99.7%) suitable for analytical and biological experiments.
  • Molecular weight 297.27 g·mol⁻1; formula C11H15N5O5.
  • Soluble in DMSO (130 mg/mL); sparingly soluble in water (2 mg/mL); ultrasonic or warming recommended to aid dissolution.
  • Stable as a powder at -20°C for extended storage.
  • Available in small research quantities (for example, 5 mg) for laboratory use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.