Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.2% | 305.29 | C13H15N5O4 | 1 ML
SDP

Catalog No. 5000203415
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is an alkyne-functionalized deoxyguanosine used as a click-chemistry reagent for biochemical and molecular biology research. It provides an alkyne handle at the 6-O position for bioorthogonal conjugation reactions, and is supplied as solutions or solids for flexible use.

  • Contains an alkyne functional group for click chemistry applications.
  • Available as 10 mM solution in DMSO (1 mL) or as solid quantities (mg scale).
  • Molecular formula C13H15N5O4 and molecular weight 305.29 g/mol.
  • High purity suitable for research use (manufacturer lists ~99% purity).
  • Provided with documentation for research and handling.

Catalog No. 50-002-03415 Supplier Medchemexpress LLC Supplier No. HY15168810MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-propynyl-2'-deoxyguanosine is an alkyne-functionalized deoxyguanosine used as a click-chemistry reagent for biochemical and molecular biology research. It provides an alkyne handle at the 6-O position for bioorthogonal conjugation reactions, and is supplied as solutions or solids for flexible use.

  • Contains an alkyne functional group for click chemistry applications.
  • Available as 10 mM solution in DMSO (1 mL) or as solid quantities (mg scale).
  • Molecular formula C13H15N5O4 and molecular weight 305.29 g/mol.
  • High purity suitable for research use (manufacturer lists ~99% purity).
  • Provided with documentation for research and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.