Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.2% | 305.29 | C13H15N5O4 | 25 MG
SDP

Catalog No. 5000201978
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is an alkyne-modified deoxyguanosine used as a click chemistry reagent for bioorthogonal labeling and conjugation in biochemical research. It is supplied as a solid for synthetic and labeling workflows and is suitable for copper-catalyzed azide-alkyne cycloaddition applications.

  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Molecular formula C13H15N5O4; molecular weight 305.29.
  • High purity for research applications (approximately 99.2%).
  • Stable as a powder at -20°C for long-term storage.
  • Supplied in small milligram quantities for synthetic and labeling workflows.

Catalog No. 50-002-01978 Supplier Medchemexpress LLC Supplier No. HY15168825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-propynyl-2'-deoxyguanosine is an alkyne-modified deoxyguanosine used as a click chemistry reagent for bioorthogonal labeling and conjugation in biochemical research. It is supplied as a solid for synthetic and labeling workflows and is suitable for copper-catalyzed azide-alkyne cycloaddition applications.

  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition.
  • Molecular formula C13H15N5O4; molecular weight 305.29.
  • High purity for research applications (approximately 99.2%).
  • Stable as a powder at -20°C for long-term storage.
  • Supplied in small milligram quantities for synthetic and labeling workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.