Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.2% | 305.29 | C13H15N5O4 | 50 MG
SDP

Catalog No. 5000202297
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is an alkyne-functionalized nucleoside used as a click chemistry reagent for biochemical research and labeling applications.

  • Molecular formula: C13H15N5O4.
  • Molecular weight: 305.29.
  • Cas number: 1640051-47-3.
  • Purity: 99.19% (data sheet; batch values may vary).
  • Form: solid; appearance white to light yellow.
  • Storage: powder at -20°C for long term, 4°C for shorter storage; in solvent store at -80°C for extended periods.
  • Pack size: 50 mg.

Catalog No. 50-002-02297 Supplier Medchemexpress LLC Supplier No. HY15168850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-propynyl-2'-deoxyguanosine is an alkyne-functionalized nucleoside used as a click chemistry reagent for biochemical research and labeling applications.

  • Molecular formula: C13H15N5O4.
  • Molecular weight: 305.29.
  • Cas number: 1640051-47-3.
  • Purity: 99.19% (data sheet; batch values may vary).
  • Form: solid; appearance white to light yellow.
  • Storage: powder at -20°C for long term, 4°C for shorter storage; in solvent store at -80°C for extended periods.
  • Pack size: 50 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.