Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.2% | 305.29 g/mol | C13H15N5O4 | 5 MG
SDP

Catalog No. 5000202667
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is a nucleoside derivative used as a click chemistry reagent; it contains a terminal alkyne at the 6-O position of 2'-deoxyguanosine and is supplied for biochemical research applications.

  • Contains a terminal alkyne suitable for click chemistry.
  • Enables bioorthogonal labeling and conjugation of nucleosides.
  • Available as solid quantities and as a 10 mM solution in DMSO for immediate use.
  • High reported purity (≈99.2%).
  • Molecular weight 305.29 g/mol; formula C13H15N5O4.

Catalog No. 50-002-02667 Supplier Medchemexpress LLC Supplier No. HY1516885MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-propynyl-2'-deoxyguanosine is a nucleoside derivative used as a click chemistry reagent; it contains a terminal alkyne at the 6-O position of 2'-deoxyguanosine and is supplied for biochemical research applications.

  • Contains a terminal alkyne suitable for click chemistry.
  • Enables bioorthogonal labeling and conjugation of nucleosides.
  • Available as solid quantities and as a 10 mM solution in DMSO for immediate use.
  • High reported purity (≈99.2%).
  • Molecular weight 305.29 g/mol; formula C13H15N5O4.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.