Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.5% | 305.29 g/mol | C13H15N5O4 | 10 MG
SDP

Catalog No. 5000202068
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is a modified nucleoside containing an alkyne (propynyl) group at the 6-O position of 2'-deoxyguanosine. It is used as a click-chemistry reagent for biochemical labeling and conjugation in research applications.

  • Cas number: 1640051-47-3.
  • Chemical formula: C13H15N5O4.
  • Molecular weight: 305.29 g/mol.
  • Purity: 99.51% (reported by distributors).
  • Available formats: powder masses (e.g., 5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and solution (10 mM in 1 mL) reported.
  • Storage: store powder at -20 °C for long term; short-term refrigerated storage also reported.
  • Intended use: for research use only; biochemical labeling and click-chemistry applications.

Catalog No. 50-002-02068 Supplier Medchemexpress LLC Supplier No. HY15168810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

6-O-propynyl-2'-deoxyguanosine is a modified nucleoside containing an alkyne (propynyl) group at the 6-O position of 2'-deoxyguanosine. It is used as a click-chemistry reagent for biochemical labeling and conjugation in research applications.

  • Cas number: 1640051-47-3.
  • Chemical formula: C13H15N5O4.
  • Molecular weight: 305.29 g/mol.
  • Purity: 99.51% (reported by distributors).
  • Available formats: powder masses (e.g., 5 mg, 10 mg, 25 mg, 50 mg, 100 mg) and solution (10 mM in 1 mL) reported.
  • Storage: store powder at -20 °C for long term; short-term refrigerated storage also reported.
  • Intended use: for research use only; biochemical labeling and click-chemistry applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.