Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 6-O-propynyl-2'-deoxyguanosine | 1640051-47-3 | 99.5% | 305.29 g/mol | C13H15N5O4 | 100 MG
SDP

Catalog No. 5000203417
Encompass_Preferred

6-O-propynyl-2'-deoxyguanosine is an alkyne-containing nucleoside used as a click-chemistry reagent for biochemical and nucleic acid research. It provides an alkyne handle for conjugation and labeling in vitro applications.

  • Alkyne-containing nucleoside for click chemistry applications.
  • Purity approximately 99.5% by HPLC.
  • CAS 1640051-47-3; formula C13H15N5O4; MW 305.29 g/mol.
  • Appearance: white to light yellow solid.
  • Soluble in DMSO (100 mg/mL); ultrasonic may be required.
  • Storage recommendations: powder -20°C for long term; in solvent -80°C for short term.
  • Pack size: 100 mg.

Catalog No. 50-002-03417 Supplier Medchemexpress LLC Supplier No. HY151688100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-O-propynyl-2'-deoxyguanosine is an alkyne-containing nucleoside used as a click-chemistry reagent for biochemical and nucleic acid research. It provides an alkyne handle for conjugation and labeling in vitro applications.

  • Alkyne-containing nucleoside for click chemistry applications.
  • Purity approximately 99.5% by HPLC.
  • CAS 1640051-47-3; formula C13H15N5O4; MW 305.29 g/mol.
  • Appearance: white to light yellow solid.
  • Soluble in DMSO (100 mg/mL); ultrasonic may be required.
  • Storage recommendations: powder -20°C for long term; in solvent -80°C for short term.
  • Pack size: 100 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.