Learn More
Cayman Chemical 6-O-Sulfo-β-cyclodextrin (sodium salt), Purity: ≥95%, CAS: 197587-31-8, 50 mg
SDP
Catalog No. 502988580
Encompass_Preferred
An anionic cyclodextrin; has been used for the chiral separation of nadolol racemates by capillary electrophoresis
Catalog No. 50-298-8580 Supplier Cayman Chemical Supplier No. 3016850 MG
An anionic cyclodextrin; has been used for the chiral separation of nadolol racemates by capillary electrophoresis