Learn More
MSC SPI Digital Protractor Inclinometer, 0.10 deg, 360 deg, 1 ea
Catalog No. 502241528
Encompass
This digital combination protractor and inclinometer provides precise angle measurements up to 360 degrees. It features a resolution of 0.10 degrees and is designed for applications requiring accurate level and plumb readings.
  • Provides 0.10 degree resolution for precise readings
  • Offers 360 degree angle measurement range
  • Features accuracy of 0.1 degrees at level 10° and plumb 10°, with a maximum error of 0.2 degrees
  • Includes digital combination style for versatile use
  • Lacks a magnetic base
  • Does not have data output capability
This item is not returnable. View return policy
Catalog No. 50-224-1528 Supplier MSC Supplier No. 80509904
This digital combination protractor and inclinometer provides precise angle measurements up to 360 degrees. It features a resolution of 0.10 degrees and is designed for applications requiring accurate level and plumb readings.
  • Provides 0.10 degree resolution for precise readings
  • Offers 360 degree angle measurement range
  • Features accuracy of 0.1 degrees at level 10° and plumb 10°, with a maximum error of 0.2 degrees
  • Includes digital combination style for versatile use
  • Lacks a magnetic base
  • Does not have data output capability