Learn More
Apexbio Technology LLC 6-Hydroxydopamine hydrobromide(Synonyms: Oxidopamine hydrobromide, 6-OHDA hydrobromide, 6-Hydroxydopamine HBr), 50mg, CAS: 636-00-0.
SDP
Catalog No. 5000061286
Encompass_Preferred
6-Hydroxydopamine hydrobromide (CAS 636-00-0) commonly referred to as 6-OHDA hydrobromide is a catecholamine-derived neurotoxin frequently used in neuroscience research to induce selective dopaminergic neuronal degeneration Upon administration it generates intracellular reactive oxygen species (ROS) activating apoptotic pathways via caspase-3 -8 and -9 as demonstrated in PC12 cell studies In rodent models targeted injection into substantia nigra striatum or medial forebrain bundle leads to loss of tyrosine hydroxylase-positive neurons and resultant locomotor dysfunction resembling symptoms observed in Parkinson s disease Consequently it serves as a valuable tool for elucidating dopaminergic dysfunction mechanisms and evaluating therapeutic candidates
Catalog No. 50-000-61286 Supplier Apexbio Technology LLC Supplier No. B709950
6-Hydroxydopamine hydrobromide (CAS 636-00-0) commonly referred to as 6-OHDA hydrobromide is a catecholamine-derived neurotoxin frequently used in neuroscience research to induce selective dopaminergic neuronal degeneration Upon administration it generates intracellular reactive oxygen species (ROS) activating apoptotic pathways via caspase-3 -8 and -9 as demonstrated in PC12 cell studies In rodent models targeted injection into substantia nigra striatum or medial forebrain bundle leads to loss of tyrosine hydroxylase-positive neurons and resultant locomotor dysfunction resembling symptoms observed in Parkinson s disease Consequently it serves as a valuable tool for elucidating dopaminergic dysfunction mechanisms and evaluating therapeutic candidates