Learn More
Cayman Chemical 6-OHDA (hydrobromide), Purity: ≥95%, CAS: 636-00-0, 50 mg
SDP
Catalog No. 502932599
Encompass_Preferred
A catecholaminergic neurotoxin; has been used to induce dopaminergic lesions and parkinsonian symptoms as a model of Parkinson's disease in rodents; induces locomotor impairment in mice and loss of tyrosine hydroxylase-reactive neurons in mouse brain when administered into the substantia nigra, MFB, or striatum at doses of 9, 6, and 18 μg, respectively; decreases dopamine and norepinephrine levels in rat brain at 200 μg; induces apoptosis in primary rat cortical cells and PC12 rat adrenal cells at 50 μM; activates caspase-3, -8, and -9 in a time-dependent manner and increases the level of intracellular ROS in PC12 cells at 75 μM; induces apoptosis in rat substantia nigra when stereotaxically injected into the right MFB
Catalog No. 50-293-2599 Supplier Cayman Chemical Supplier No. 2533050 MG
A catecholaminergic neurotoxin; has been used to induce dopaminergic lesions and parkinsonian symptoms as a model of Parkinson's disease in rodents; induces locomotor impairment in mice and loss of tyrosine hydroxylase-reactive neurons in mouse brain when administered into the substantia nigra, MFB, or striatum at doses of 9, 6, and 18 μg, respectively; decreases dopamine and norepinephrine levels in rat brain at 200 μg; induces apoptosis in primary rat cortical cells and PC12 rat adrenal cells at 50 μM; activates caspase-3, -8, and -9 in a time-dependent manner and increases the level of intracellular ROS in PC12 cells at 75 μM; induces apoptosis in rat substantia nigra when stereotaxically injected into the right MFB