Learn More
Medchemexpress LLC 6-Oxohexyl 2-hexyldecanoate | 1849616-54-1 | 80.0% | C22H42O3 | 250 MG
SDP
Catalog No. 5000352833
Encompass_Preferred
6-Oxohexyl 2-hexyldecanoate, 80% (ELSD) is an intermediate used in the synthesis of ALC-0315. It can be utilized in research related to drug delivery, including mRNA delivery. ALC-0315 is an ionizable amino lipid and a key component of LNP delivery vehicles.
  • Intermediate in the synthesis of ALC-0315
  • Suitable for research on drug delivery
  • Supports mRNA delivery studies
  • Contributes to LNP delivery vehicles
Catalog No. 50-003-52833 Supplier Medchemexpress LLC Supplier No. 250MGHY142977
6-Oxohexyl 2-hexyldecanoate, 80% (ELSD) is an intermediate used in the synthesis of ALC-0315. It can be utilized in research related to drug delivery, including mRNA delivery. ALC-0315 is an ionizable amino lipid and a key component of LNP delivery vehicles.
  • Intermediate in the synthesis of ALC-0315
  • Suitable for research on drug delivery
  • Supports mRNA delivery studies
  • Contributes to LNP delivery vehicles