Learn More
Medchemexpress LLC 6-oxolithocholic acid | 2393-61-5 | MFCD00070044 | 100.0% | 390.56 | C24H38O4 | 10 MG
SDP
Catalog No. 5000284608
Encompass_Preferred
6-Oxolithocholic acid is an endogenous bile acid metabolite used as a research reagent to study bile acid metabolism, cytotoxicity, and related cellular processes. It is supplied as a high-purity solid or solution suitable for biochemical and cell biology assays and should be stored under recommended conditions.
Catalog No. 50-002-84608 Supplier Medchemexpress LLC Supplier No. HYW58753010MG
6-Oxolithocholic acid is an endogenous bile acid metabolite used as a research reagent to study bile acid metabolism, cytotoxicity, and related cellular processes. It is supplied as a high-purity solid or solution suitable for biochemical and cell biology assays and should be stored under recommended conditions.