Learn More
Burch Bottle & Packaging Inc 6 OZ GLASS STRAIGHT-SIDE 12/CS
Catalog No. NC1778369
Encompass
Stock number 1SS061GHG is a 6oz Straight Sided Jar with a 63/400 continuous thread finish. Straight Sided Jars have a simple but elegant look. The wide mouth of the 6oz Straight Sided is perfect for scooping or pouring. The large label panel gives you plenty of space for a creative label, while the round shape makes for easy labeling.
Catalog No. NC1778369 Supplier Burch Bottle & Packaging Inc Supplier No. 1SS061GHG
Stock number 1SS061GHG is a 6oz Straight Sided Jar with a 63/400 continuous thread finish. Straight Sided Jars have a simple but elegant look. The wide mouth of the 6oz Straight Sided is perfect for scooping or pouring. The large label panel gives you plenty of space for a creative label, while the round shape makes for easy labeling.