Learn More
JANT PHARMACAL CORPORATION Accutest ValuPak Drug Test Cup-6. AMP1000, COC 300, MET 1000, OPI 2000, PCP 25, THC 50 25 Tests/Kit Box
SDP
Catalog No. NC3457081
Encompass_Preferred
Accutest CLIA Waived 6 panel drug test cup is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.
Catalog No. NC3457081 Supplier JANT PHARMACAL CORPORATION Supplier No. DS487
Accutest CLIA Waived 6 panel drug test cup is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.