Learn More
Cliawaived Inc IDTC-II 6 Panel Instant Drug Test Cards
SDP
Catalog No. NC1859235
Encompass_Preferred
(25 Tests/Box)Tests for AMP500 / BZO300 / COC150 / MET500 / OPI (MOR 300) / THC50The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.
This item is not returnable. View return policy
Catalog No. NC1859235 Supplier Cliawaived Inc Supplier No. CLIAIDTCII06264
(25 Tests/Box)Tests for AMP500 / BZO300 / COC150 / MET500 / OPI (MOR 300) / THC50The newly designed IDTC-II dip card features a hermetic seal to prevent the card from flooding. The device features a flat and separate cap so it can be easily photographed without contamination.