Learn More
Spherotech 6 PEAK ULTRA RAINBOW CALIB 3ML
Catalog No. NC9207381
Encompass_Preferred
UltraRainbow Calibration Particle Kit, 5.0-5.4 um, 2E6/mL, 1x3 mL (blank) & 1E7/mL, 1x2 mL (mix)
Catalog No. NC9207381 Supplier Spherotech Supplier No. URCP502K
UltraRainbow Calibration Particle Kit, 5.0-5.4 um, 2E6/mL, 1x3 mL (blank) & 1E7/mL, 1x2 mL (mix)