Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Phenoxynicotinic acid, 97%, Thermo Scientific™

Catalog No. CC19601DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC19601DE 5 g
CC19601CB 250 mg
CC19601DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC19601DE Supplier Thermo Scientific Chemicals Supplier No. CC19601DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 51362-38-0
Molecular Formula C12H9NO3
Molecular Weight (g/mol) 215.208
MDL Number MFCD02682068
InChI Key GFEUNYLQJDQNAN-UHFFFAOYSA-N
Synonym 6-phenoxynicotinic acid, 3-pyridinecarboxylic acid, 6-phenoxy, 6-phenoxynicotinicacid, 6-phenoxy-nicotinic acid, 2-phenoxypyridine-5-carboxylic acid, 3-pyridinecarboxylicacid, 6-phenoxy, 6-phenyloxy-3-pyridinecarboxylic acid
PubChem CID 2776497
IUPAC Name 6-phenoxypyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)OC2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 215.21
Percent Purity 97%
Chemical Name or Material 6-Phenoxynicotinic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.