Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Phenyl-2-pyridone 98.0+%, TCI America™
SDP

Catalog No. P09481G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09481G 1 g
P09485G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P09481G Supplier TCI America Supplier No. P09481G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19006-82-7
Molecular Formula C11H9NO
Molecular Weight (g/mol) 171.20
MDL Number MFCD00059760
InChI Key ZLUIDXRFWSPPCC-UHFFFAOYSA-N
PubChem CID 87889
IUPAC Name 6-phenyl-1,2-dihydropyridin-2-one
SMILES O=C1NC(=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 197°C
Color Yellow
Quantity 1 g
Formula Weight 171.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Phenyl-2-pyridone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.