Learn More
Medchemexpress LLC 6-phenylimidazo[2,1-b]thiadiazole-2-sulfonamide | 63735-71-7 | MFCD02064987 | 99.8% | 280.33 g·mol⁻1 | C10H8N4O2S2 | 5 MG
SDP
Catalog No. 5000240031
Encompass_Preferred
AEG3482 is a small-molecule JNK inhibitor used in research to probe apoptosis and stress-response signaling. It binds Hsp90, promotes HSF1-dependent induction of heat shock proteins such as Hsp70 and Hsp25, and shows antiapoptotic activity in cellular and in vivo models. Supplied as a solid for laboratory use, it is intended for research only.
Catalog No. 50-002-40031 Supplier Medchemexpress LLC Supplier No. HY1075995MG
AEG3482 is a small-molecule JNK inhibitor used in research to probe apoptosis and stress-response signaling. It binds Hsp90, promotes HSF1-dependent induction of heat shock proteins such as Hsp70 and Hsp25, and shows antiapoptotic activity in cellular and in vivo models. Supplied as a solid for laboratory use, it is intended for research only.