Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Phenylnicotinic acid, 95%, Thermo Scientific™

Catalog No. CC21101CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC21101CB 250 mg Amber Glass Bottle
CC21101DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC21101CB Supplier Thermo Scientific Chemicals Supplier No. CC21101CB
Add to Cart

Chemical Identifiers

CAS 29051-44-3
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD03086128
InChI Key DLFLQXUYRFIFOK-UHFFFAOYSA-N
Synonym 6-phenylnicotinic acid, 3-pyridinecarboxylic acid, 6-phenyl, 6-phenyl-nicotinic acid, 2-phenyl pyridine-5-carboxylic acid, 6-phenyl nicotinic acid, acmc-1cmxf, 2-phenyl-5-carboxypyridine, 3-carboxy-6-phenylpyridine, 2-phenyl-5-pyridinecarboxylic acid, 6-phenyl-3-pyridinecarboxylic acid
PubChem CID 120118
IUPAC Name 6-phenylpyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)C2=NC=C(C=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 6-Phenylnicotinic acid
Hazard Category Warning
Hazard Statement GHS P Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.