Learn More
Medchemexpress LLC 6-Phosphogluconic acid trisodium | 53411-70-4 | 99.8% | 342.08 | 10 MG
SDP
Catalog No. 5000453578
Encompass_Preferred
6-Phosphogluconic acid trisodium is a potent and competitive phosphoglucose isomerase (PGI) inhibitor with Kis of 48 μM for glucose 6-phosphate and 42 μM for fructose 6-phosphate. It is also an endogenous metabolite.
  • Potent and competitive phosphoglucose isomerase (PGI) inhibitor
  • Endogenous metabolite
Catalog No. 50-004-53578 Supplier Medchemexpress LLC Supplier No. HYW01118410MG
6-Phosphogluconic acid trisodium is a potent and competitive phosphoglucose isomerase (PGI) inhibitor with Kis of 48 μM for glucose 6-phosphate and 42 μM for fructose 6-phosphate. It is also an endogenous metabolite.
  • Potent and competitive phosphoglucose isomerase (PGI) inhibitor
  • Endogenous metabolite