Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-piperidin-1-ylnicotinaldehyde, Thermo Scientific™

Catalog No. MO07199DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO07199DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO07199DA Supplier Thermo Scientific Chemicals Supplier No. MO07199DA
Add to Cart

Chemical Identifiers

CAS 241816-11-5
Molecular Formula C11H14N2O
Molecular Weight (g/mol) 190.246
InChI Key RKIMIISNVCLRLA-UHFFFAOYSA-N
Synonym 6-piperidin-1-ylnicotinaldehyde, 6-piperidin-1-yl nicotinaldehyde, 6-piperidin-1-yl pyridine-3-carbaldehyde, 6-piperidinonicotinaldehyde, 6-1-piperidinyl nicotinaldehyde, 6-1-piperidinyl pyridine-3-carboxaldehyde, 6-piperidylpyridine-3-carbaldehyde, acmc-20alxp, 3-pyridinecarboxaldehyde,6-1-piperidinyl, 3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-5'-carbaldehyde
PubChem CID 2763980
IUPAC Name 6-piperidin-1-ylpyridine-3-carbaldehyde
SMILES C1CCN(CC1)C2=NC=C(C=C2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 190.24
Percent Purity 97%
Chemical Name or Material 6-Piperidin-1-ylnicotinaldehyde
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.